Merck
  • [Gas-chromatographic method of determining the levels of tri-allate in the water, soil and plants].
[Gas-chromatographic method of determining the levels of tri-allate in the water, soil and plants].
Gigiena i sanitariia (1989-05-01)
I I Pilenkova, R G Iurkova, A D Fa'tianova
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Triallat, PESTANAL®, analytical standard