Merck
Biological N-oxidation of piperidine in vitro.
Zhongguo yao li xue bao = Acta pharmacologica Sinica (1989-05-01)
D Y Wang, J W Gorrod, A H Beckett
ZUSAMMENFASSUNG
The biological N-oxidation of piperidine, a pharmacologically active biogenic amine of mammals and human beings, was studied in vitro. After incubation of piperidine-HCl in a fortified rat liver microsomal preparation (9000 x g supernatant) at 37 degrees C for 30 min, 2 metabolites were detected. They were identified as N-hydroxy piperidine and 2, 3, 4, 5-tetrahydro-pyridine-1-oxide as evidenced by TLC, GLC, HPLC, GC-MS and MS.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1-Hydroxypiperidin, ≥96.0% (GC)