Merck
[3H]Ro 19-6327: a reversible ligand and affinity labelling probe for monoamine oxidase-B.
European journal of pharmacology (1989-03-29)
A M Cesura, M D Galva, R Imhof, E Kyburz, G B Picotti, M Da Prada
ZUSAMMENFASSUNG
This study demonstrated the existence of specific binding sites for [3H]Ro 19-6327 in human platelet membranes. This compound is a novel, time-dependent inhibitor of monoamine oxidase type B (MAO-B) and is structurally closely related to [3H]Ro 16-6491. The density of the sites labelled with high affinity by [3H]Ro 19-6327 was similar to that observed in previous studies with [3H]Ro 16-6491 as ligand. Binding was reversible at 20 degrees C and showed a relatively slow dissociation (t1/2 = 220 min). The dissociation rate was markedly decreased (t1/2 = greater than 24h) at 0 degrees C. MAO-B, but not MAO-A inhibitors, effectively prevented the binding of [3H]Ro 19-6327. Like [3H]Ro 16-6491, [3H]Ro 19-6327 is recognized as a substrate by MAO-B, being eventually deaminated by the enzyme. Since the deaminated aldehyde derivative of Ro 19-6327 did not inhibit MAO-B, a still unidentified reversible adduct, formed at the MAO-B active site, might explain the high potency and selectivity of [3H]Ro 19-6327. Incubation of the radioligand-enzyme complex from platelet and brain membranes with NaBH3CN and acetic acid (to pH 4.5) caused the irreversible incorporation of the radioactivity into a single polypeptide as shown by SDS-PAGE analysis. This polypeptide had a molecular weight identical to that of the MAO-B subunit, i.e. 58,000. The presence of unlabelled MAO-B inhibitors in the incubation mixture prevented the covalent incorporation of [3H]Ro 19-6327. The irreversible MAO-B inhibitor, [3H] pargyline, labelled a protein with a molecular weight identical to the protein labelled by [3H]Ro 19-6327.(ABSTRACT TRUNCATED AT 250 WORDS)