Merck
  • Luteinizing hormone-releasing hormone enhances polyphosphoinositide breakdown in rat granulosa cells.
Luteinizing hormone-releasing hormone enhances polyphosphoinositide breakdown in rat granulosa cells.
Biochemical and biophysical research communications (1985-08-15)
F Ma, P C Leung
ZUSAMMENFASSUNG
A 2-min addition of LHRH to [3H]inositol-prelabeled rat granulosa cells in primary culture evoked significant increases in the accumulation of [3H]inositol phosphates, i.e. radiolabeled inositol monophosphate (IP), inositol diphosphate (IP2), and inositol triphosphate (IP3) levels increased to 210, 590 and 520%, respectively, when compared to control cultures. By contrast, addition of FSH failed to elicit such a response. The effect of LHRH was completely blocked by the concomitant presence of a specific LHRH antagonist. LHRH evoked increase in [3H]IP3 and [3H]IP2 accumulation as early as 30 sec, while the increase in [3H]IP became significant at 2 min. These data support the hypothesis that polyphosphoinositide breakdown may be an early step in the intracellular signal mechanism which mediates the action of LHRH.