Merck
  • Actions of synthetic leukotrienes on platelets and blood vessels in the anesthetised pig: the release of a platelet derived vasodilator.
Actions of synthetic leukotrienes on platelets and blood vessels in the anesthetised pig: the release of a platelet derived vasodilator.
Prostaglandins (1985-06-01)
L G Letts, M Cirino, P Yusko, B Fitzsimmons, A W Ford-Hutchinson, J Rokach
ZUSAMMENFASSUNG
The actions of leukotrienes (LT's) C4, D4, E4 and F4 have been investigated in the perfused hind-limb of the anesthetized pig. In the blood perfused hind limb LTC4, D4 and E4 increased the perfusion pressure in a dose-dependent fashion whereas LTF4 decreased perfusion pressure. In the Tyrode perfused hind limb all LT's increased perfusion pressure (rank order potency LTC4 = LTD4 much greater than LTF4). The actions of LTF4 were not affected by a wide variety of pharmacological treatments, including indomethacin, methysergide and FPL-55712. The LT's aggregated porcine platelets (rank order potency LTC4 greater than LTF4 greater than LTD4) and induced the release of a platelet-derived vasodilatory mediator. The results provide pharmacological evidence of specific leukotriene receptors in vivo and that leukotrienes can independently modulate blood flow. These data suggest that important interactions may occur between platelets, the arachidonate lipoxygenase products and platelet-derived substances in response to inflammatory stimuli in the cardiovascular system.