Merck
  • Estriol and its conjugates in late pregnancy determined by extraction with Carbopack B and liquid chromatography with fluorometric detection.
Estriol and its conjugates in late pregnancy determined by extraction with Carbopack B and liquid chromatography with fluorometric detection.
Clinical chemistry (1985-10-01)
F Andreolini, C Borra, F Caccamo, A Di Corcia, I Nicoletti, R Samperi, F Improta
ZUSAMMENFASSUNG
We report a method for measuring estriol and its intact conjugates in urine, serum, and amniotic fluid. A single assay can be done within about 50 min, eight samples assayed in less than 5 h. A 70-microL urine sample is diluted and the estriol conjugates are adsorbed from it onto graphitized carbon black (Carbopack B, Supelco). After two washings, the analytes are desorbed with chloroform/methanol (60/40, by vol) containing tetrapropylammonium bromide. After solvent evaporation, the residue is redissolved in 100 microL of water/acetonitrile and 20 microL is injected into the chromatograph. Or 1 mL of serum or 0.5 mL of amniotic fluid is deproteinized with cold methanol, then passed through the Carbopack column. After three washings, the estriol and its conjugates are desorbed and treated as for urine. Mean analytical recoveries of the analytes in any of these body fluids were within about 92-98%, except for estriol-3-sulfate-16 alpha-glucuronide in serum (mean recovery 88.3%). The limit of sensitivity is well below the concentrations of clinical interest, and the method is not susceptible to substantial interferences.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Progesteron, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesteron, ≥99%
Sigma-Aldrich
Progesteron, meets USP testing specifications
Sigma-Aldrich
Progesteron, γ-irradiated, BioXtra, suitable for cell culture
Supelco
FLM Carbopack X TD-Röhrchen aus deaktiviertem Edelstahl, preconditioned, pkg of 10 tubes, O.D. × L 1/4 in. (6.35 mm) × 3 1/2 in. (89 mm)
Supelco
Progesteron, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Carbopack Adsorbens, matrix Carbopack B, 60-80 mesh, bottle of 10 g
Sigma-Aldrich
Estriol, ≥97%
Supelco
Progesterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Carbopack Adsorbens, matrix Carbopack C, 80-100 mesh, bottle of 10 g
Supelco
Carbopack Adsorbens, matrix Carbopack C, 60-80 mesh, bottle of 10 g
Sigma-Aldrich
D-erythro-Dihydrosphingosine, ≥98%
Supelco
Carbopack Adsorbens, matrix Carbopack X, 40-60 mesh, bottle of 10 g
Supelco
Carbopack Adsorbens, matrix Carbopack X, 60-80 mesh, bottle of 10 g
Supelco
Estriol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Estriol 16α-(β-D-Glucuronid), ≥97%
Supelco
Progesteron, VETRANAL®, analytical standard
Supelco
Carbopack B, stainless steel TD tube, O.D. × L 1/4 × 3 1/2, unconditioned, pkg of 10 ea
Sigma-Aldrich
Estriol-3-Sulfat Natriumsalz, ≥98% (TLC)
Sigma-Aldrich
Estriol, meets USP testing specifications
Supelco
Carbopack X, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
Supelco
Carbopack B, O.D. × L 1/4 in. × 3 1/2 in., glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
Supelco
Carbopack B, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., unconditioned, pkg of 10 ea, matrix Carbopack
Supelco
Carbopack B, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea