- High-performance liquid chromatographic determination of dimethyldithiocarbamate residues in some agricultural products.
High-performance liquid chromatographic determination of dimethyldithiocarbamate residues in some agricultural products.
Journal of chromatography (1986-02-28)
E Brandsteterová, J Lehotay, O Liska, J Garaj
PMID3009509
ZUSAMMENFASSUNG
Dimethyldithiocarbamates are widely used in agriculture as active fungicides. The degradation of dimethyldithiocarbamates (ferbam, thiram) confirmed the fact that they are not stable and decompose very rapidly. The aim of this work was to apply the results obtained in high-performance liquid chromatographic quantitative analysis of residues of dithiocarbamate fungicides in some agricultural products (strawberries, maize, tobacco). The developed method enables very simple control analysis of low concentrations of dimethyldithiocarbamate residues in very short time. All limits of detection correspond with the criteria of FAO (Codex Alimentarius).