Merck
  • Potent inhibition of superoxide anion generation by PGE1 and the PGE1 analogue OP-1206 in human PMN's--unrelated to its antiplatelet PGI2-like activity.
Potent inhibition of superoxide anion generation by PGE1 and the PGE1 analogue OP-1206 in human PMN's--unrelated to its antiplatelet PGI2-like activity.
VASA. Supplementum (1987-01-01)
K Schrör, G Hecker
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Limaprost, ≥99%, crystalline