- Myasthenia-like syndrome induced by cardiovascular agents. Report of a case.
Myasthenia-like syndrome induced by cardiovascular agents. Report of a case.
Italian journal of neurological sciences (1987-04-01)
B Fierro, M G Castiglione, G Salemi, G Savettieri
PMID3036745
ZUSAMMENFASSUNG
The case of a myasthenia-like syndrome induced by cardiovascular drugs is reported. The clinical and electrophysiological features of the case are discussed.