Merck
  • Evidence suggesting a role for sperm metalloendoprotease activity in penetration of zona-free hamster eggs by human sperm.
Evidence suggesting a role for sperm metalloendoprotease activity in penetration of zona-free hamster eggs by human sperm.
The Journal of experimental zoology (1988-11-01)
E Díaz-Pérez, P Thomas, S Meizel
ZUSAMMENFASSUNG
It has been reported that metalloendoprotease (MEP) activity is involved in somatic cell membrane fusion events and in the sea urchin sperm acrosome reaction (AR). MEP activity also has been demonstrated in human and other mammalian sperm. The present study was concerned with investigating whether a human sperm MEP is important in membrane events necessary for sperm egg fusion. Ejaculated human sperm were washed, capacitated in vitro, and preincubated with the competitive MEP inhibitors phosphoramidon (50 microM) or CBZ-L-phenylalanine (1 mM), with 100 microM diethylenetriaminepentaacetic acid (DTPA), a heavy metal chelator, or as controls, with the appropriate solvents. The AR was initiated in vitro with preovulatory human follicular fluid and the sperm washed to dilute inhibitors and then coincubated with zona-free golden hamster eggs (zonae and cumuli removed with trypsin and hyaluronidase, respectively). Eggs were washed after 0.5 h, and the number of sperm remaining bound was counted. After 2.5 h further incubation, the eggs were stained with acetolacmoid or acetoorcein and penetration was assayed by counting the number of decondensed sperm heads per egg (penetration index) and the percent of penetrated eggs. The inhibitor treatments did not decrease the percentage of penetrated eggs (range 80-90%), but a significant reduction in the penetration index was observed. Phosphoramidon reduced the penetration index by 45%, CBZ-L-phenylalanine by 57%, and DTPA by 56%. None of the inhibitors decreased the penetration index or the percentage of penetrated eggs when added directly to suspensions of acrosome-reacted sperm and zona-free eggs at the diluted levels that would have been present after washing inhibitor-treated sperm.(ABSTRACT TRUNCATED AT 250 WORDS)