Merck
  • Resolution of diastereomers of N,N,N',N'-tetrakis(2-hydroxypropyl)ethylenediamine (quadrol) by reversed-phase high-performance liquid chromatography.
Resolution of diastereomers of N,N,N',N'-tetrakis(2-hydroxypropyl)ethylenediamine (quadrol) by reversed-phase high-performance liquid chromatography.
Journal of chromatography (1988-11-25)
J A Fulton, H K Pokharna, M J Dunphy, D J Smith
ZUSAMMENFASSUNG
Quadrol, N,N,N',N'-tetrakis(2-hydroxypropyl)ethylenediamine, has been recently observed to display biological activity. It is an immunostimulant and has been implicated as a potentially useful agent in accelerated wound healing. Quadrol exists as a mixture of four unique diastereomers, each of which may, upon further investigation, display differences in biological activity. This paper describes an high-performance liquid chromatographic procedure (both analytical and prep) for the separation of the Quadrol diastereomers. Gas-liquid chromatography and NMR data are presented which corroborate the high-performance liquid chromatographic results. This procedure may be used to obtain pure Quadrol diastereomers, to monitor the progress of Quadrol synthesis from propylene oxide and ethylenediamine or to develop a quantitative assay for Quadrol diastereomers.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N,N,N′,N′-Tetrakis(2-Hydroxypropyl)ethylendiamin, 98%