- Criteria for evaluating improvement in schizophrenia in psychopharmacological research (with special reference to gamma endorphin fragments).
Criteria for evaluating improvement in schizophrenia in psychopharmacological research (with special reference to gamma endorphin fragments).
The British journal of psychiatry : the journal of mental science (1988-09-01)
R Manchanda, S R Hirsch, T R Barnes
PMID3250672
ZUSAMMENFASSUNG
A review of treatment trials with DT gamma E revealed widely discrepant results. Relevant variables were the variety of measures employed for monitoring psychotic symptoms, and the different criteria used to judge the degree of improvement. The authors suggest a uniform outcome criterion for early trials of new treatments, which would generate more consistent and comparable results between studies, and give a stronger indication of the value of the treatment under test. When the data from the various treatment trials of DT gamma E were reanalysed, applying a uniform outcome criterion of improvement of a change of 80% or more on rating-scale score, the results were more consistent than would have been suspected from the original reports.