Merck
  • The milk oligosaccharide, lacto-N-fucopentaose I, inhibits attachment of mouse blastocysts on endometrial monolayers.
The milk oligosaccharide, lacto-N-fucopentaose I, inhibits attachment of mouse blastocysts on endometrial monolayers.
Journal of reproduction and fertility (1988-05-01)
S Lindenberg, K Sundberg, S J Kimber, A Lundblad
ZUSAMMENFASSUNG
Seven oligosaccharides isolated from human milk were tested for their effect in an in-vitro model of mouse blastocyst adhesion and trophoblast outgrowth on endometrial epithelial monolayers. One compound, lacto-N-fucopentaose I (LNF I), produced a significant reduction in the percentage of attached and outgrown blastocysts after co-culture for 72 h (P less than 0.001). No significant effect of any other tested oligosaccharide was obtained.