Merck
  • [Effect of the treatment of helminth infection with Panacur (Hoechst) and Banminth (Pfizer) on the productivity of laying hens].
[Effect of the treatment of helminth infection with Panacur (Hoechst) and Banminth (Pfizer) on the productivity of laying hens].
Wiadomosci parazytologiczne (1987-01-01)
A Ramisz