Merck
  • A general practice study to compare alverine citrate with mebeverine hydrochloride in the treatment of irritable bowel syndrome.
A general practice study to compare alverine citrate with mebeverine hydrochloride in the treatment of irritable bowel syndrome.
The British journal of clinical practice (1986-07-01)
G J Tudor
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Alverin -citrat (Salz)
Alverin -citrat (Salz), European Pharmacopoeia (EP) Reference Standard