Merck
  • Identification of N-carboxyethyl gamma-aminobutyric acid in bovine brain and human cerebrospinal fluid.
Identification of N-carboxyethyl gamma-aminobutyric acid in bovine brain and human cerebrospinal fluid.
Neuroscience letters (1987-06-26)
F Fussi, F Savoldi, M Curti
ZUSAMMENFASSUNG
The di-carboxylated derivative of spermidine, N-carboxyethyl gamma-aminobutyric acid (CEGABA) has been identified in bovine brain and human cerebrospinal fluid by HPLC. This discovery strongly suggests the existence of a metabolic pathway connecting polyamines and GABA via putreanine and CEGABA through progressive oxidative deamination of the amino terminal groups in spermidine.