Merck
  • High-performance liquid chromatographic determination of urinary catecholamines by direct pre-column fluorescence derivatization with 1,2-diphenylethylenediamine.
High-performance liquid chromatographic determination of urinary catecholamines by direct pre-column fluorescence derivatization with 1,2-diphenylethylenediamine.
Journal of chromatography (1986-07-11)
H Nohta, A Mitsui, Y Ohkura
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N,N′-Diphenylethylendiamin, 98%