- [Low-molecular-weight immunocorrectors of natural origin].
[Low-molecular-weight immunocorrectors of natural origin].
Antibiotiki i meditsinskaia biotekhnologiia = Antibiotics and medical biotechnology (1985-02-01)
L P Ivanitskaia, O N Oparina, T N Stekhanova, S M Navashin
PMID3890722
MATERIALIEN