- Structural study of a new antitumor antibiotic, kazusamycin.
Structural study of a new antitumor antibiotic, kazusamycin.
The Journal of antibiotics (1985-02-01)
K Komiyama, K Okada, H Oka, S Tomisaka, T Miyano, S Funayama, I Umezawa
PMID3922934
ZUSAMMENFASSUNG
The structure of a new antitumor antibiotic, kazusamycin produced by Streptomyces sp. No. 81-484, was determined on the basis of its spectral and chemical properties. Kazusamycin has a structure characteristic of an unsaturated, branched-chain fatty acid with a terminal delta-lactone ring.