Merck
  • A benzodiazepine antagonist inhibits the cerebral metabolic and respiratory depressant effects of fentanyl.
A benzodiazepine antagonist inhibits the cerebral metabolic and respiratory depressant effects of fentanyl.
Life sciences (1985-06-10)
N Naughton, W E Hoffman, P Larscheid, J M Cook, R F Albrecht, D J Miletich
ZUSAMMENFASSUNG
It is reported that benzodiazepines such as diazepam will stimulate the opiate receptor system and that B-carboline drugs, which are benzodiazepine antagonists, may interact with opiate receptors directly. The ability of 3-hydroxymethyl-B-carboline (3-HMC) to antagonize several parameters of fentanyl anesthesia was tested here in rats. Fentanyl (25 and 100 micrograms/kg iv) produced dose dependent depression of cerebral blood flow (CBF), measured by radioactive microspheres, and cerebral oxygen consumption (CMRO2). These effects were significantly inhibited by 10 mg/kg 3-HMC iv. To test for the specificity of this effect, 3-HMC was also given to rats ventilated with inspire concentrations of 2% halothane. Halothane depressed CMRO2 equally in 3-HMC and vehicle treated rats, indicating no significant effect of the benzodiazepine antagonist. Blood pressure was increased in 3-HMC compared to vehicle treated animals during both fentanyl and halothane anesthesia. CBF was increased in 3-HMC vs vehicle treated rats during halothane anesthesia but this could be accounted for by the elevated blood pressure and lack of cerebral autoregulation rather than a direct cerebrovascular effect. 3-HMC decreased the sleep time and respiratory depressant effects of fentanyl but enhanced the analgesic effects of the opiate, as measured by time to respond to a hot plate stimulus. These results indicate that 3-HMC has the ability to specifically antagonize fentanyl anesthesia. These effects may be produced by an action of 3-HMC at the benzodiazepine receptor and/or by an action of the B-carboline at opioid receptors.