Merck
  • Effectiveness of seasonal community-based mass-expulsion chemotherapy in the control of human hookworm infections in endemic communities.
Effectiveness of seasonal community-based mass-expulsion chemotherapy in the control of human hookworm infections in endemic communities.
Public health (1985-09-01)
J K Udonsi