Merck
  • The stimulatory effect of a non-opiate beta-endorphin fragment on arginine-vasopressin release in rats.
The stimulatory effect of a non-opiate beta-endorphin fragment on arginine-vasopressin release in rats.
Acta medica Hungarica (1984-01-01)
F Laczi, J E Sykes, V M Wiegant, D de Wied
ZUSAMMENFASSUNG
The effect of the non-opiate beta-endorphin (beta E) fragment 2-9 and related peptides on immunoreactive (IR) arginine8 -vasopressin (AVP) levels was studied in the rat eye plexus plasma. Additionally, the effect of beta E 2-9 on AVP release from pituitary neurointermediate lobes ( NILs ), in vitro was studied. IR AVP levels in the eye plexus plasma increased after subcutaneous (s.c.) injection of beta E 2-9. In female rats peak values were observed 2 min after the administration of beta E 2-9 (10 micrograms/rat). Male rats were 100 times more sensitive than female rats. A dose-response study revealed a U-shaped relationship for this effect of beta E 2-9 in animals of both sexes. From structure-activity studies it appeared that beta E 2-9 was the most effective fragment. Intracerebroventricular (i.c.v.) administration of beta E 2-9 did not affect the IR-AVP levels in eye plexus plasma. Moreover, AVP release from the rat NILs cells in vitro was stimulated by perfusion with beta E 2-9, indicating a direct effect of the peptide on the pituitary. Therefore we suggest that beta E 2-9 increases AVP release by a direct action on the posterior pituitary.