Merck
  • Deuterium labelled steroid hormones: syntheses and application in quantitation and endocrinology.
Deuterium labelled steroid hormones: syntheses and application in quantitation and endocrinology.
Journal of steroid biochemistry (1981-08-01)
D W Johnson, G Phillipou, R F Seamark