- Inhibition of polymorphonuclear leukocyte functions by fluorinated nitrobenzenes.
Inhibition of polymorphonuclear leukocyte functions by fluorinated nitrobenzenes.
Chemico-biological interactions (1984-12-01)
J G Elferink, M Deierkauf
PMID6509685
ZUSAMMENFASSUNG
The bifunctional fluorinated nitrobenzenes, 1,5-difluoro-2,4-dinitrobenzene (DFDNB) and 4,4'-difluoro-3,3'-dinitrodiphenyl sulfone (DFDNDPS), and the monofunctional 1-fluoro-2,4-dinitrobenzene (FDNB) inhibit chemotaxis, phagocytosis, exocytosis and the respiratory burst of rabbit polymorphonuclear leukocytes. Inhibition occurs in the micromolar concentration range; the bifunctional compounds are stronger inhibitory than the monofunctional one. The inhibitory effect can be counteracted by sulfhydryl compounds and not with amino-group containing compounds. The results suggest that an interaction with vulnerable sulfhydryl groups, located in a hydrophobic surrounding, is the basis of the inhibitory effect of the fluorinated nitrobenzenes.