- [The effect of urea in a topical corticoid preparation on vasoconstriction].
[The effect of urea in a topical corticoid preparation on vasoconstriction].
Zeitschrift fur Hautkrankheiten (1983-08-15)
H Früh
PMID6624181
ZUSAMMENFASSUNG
It was checked by vasoconstriction tests, whether a new topical preparation containing 0.1% halcinonide in combination with 10% urea possibly caused an increase in vasoconstriction. There was no significant difference found between the preparations with and without urea.