Merck
Reduction of octoclothepine-N-oxide by rat liver microsomes.
European journal of drug metabolism and pharmacokinetics (1981-01-01)
O Helia, E Vargova, A Jindra
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Octoclothepin maleate salt, solid