Merck
  • Significance of apparent half-lives of a metabolite with a higher elimination rate than its parent drug.
Significance of apparent half-lives of a metabolite with a higher elimination rate than its parent drug.
Drug intelligence & clinical pharmacy (1982-02-01)
T B Vree, C A Hekster, E van der Kleijn
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sulfamethoxazol Unreinheit A, European Pharmacopoeia (EP) Reference Standard