Merck
N-nitrosodimethylamine in malt beverages--anticipatory action by the brewing industry.
Regulatory toxicology and pharmacology : RTP (1982-03-01)
W A Hardwick, D H Hickman, N O Jangaard, W J Ladish, M C Meilgaard
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Hordenin, ≥97.0% (HPLC)