- Immunosuppressive activities of 6(5H)-phenanthridinone, a new poly(ADP-ribose)polymerase inhibitor.
Immunosuppressive activities of 6(5H)-phenanthridinone, a new poly(ADP-ribose)polymerase inhibitor.
International journal of immunopharmacology (1995-04-01)
D Weltin, V Picard, K Aupeix, M Varin, D Oth, J Marchal, P Dufour, P Bischoff
PMID7672878
ZUSAMMENFASSUNG
6(5H)-phenanthridinone, a recently identified poly(ADP-ribose)polymerase (PARP) inhibitor, is able, at micromolar concentrations, to inhibit concanavalin A-induced lymphocyte proliferation and to potentiate the effect of gamma radiation upon murine spleen cells. When added at the onset of a mixed lymphocyte culture, this compound strongly depresses the induction of primary allogeneic (anti-H2k) cytotoxic T-lymphocytes (CTLs). Lymphokine-activated killer (LAK) induction was also found to be impaired by the PARP inhibitor. Taken together, these results clearly indicate that PARP plays a key-role in immune reactions involving cytotoxicity and that 6(5H)-phenanthridinone could be considered as a potent immunomodulator.
MATERIALIEN