Merck
  • Induction of hepatic drug-metabolizing enzymes by chlornitrofen (CNP) and CNP-amino in rats and mice.
Induction of hepatic drug-metabolizing enzymes by chlornitrofen (CNP) and CNP-amino in rats and mice.
Chemosphere (1995-04-01)
N Hanioka, K Nakano, H Jinno, M Hamamura, A Takahashi, R Yoda, T Nishimura, M Ando
ZUSAMMENFASSUNG
The induction of hepatic drug-metabolizing enzymes by chlornitrofen (CNP) and CNP-amino was studied in the liver of male rats and mice. CNP-amino increased the activities of 7-pentoxyresorufin O-depentylase (PROD) and 7-benzyloxyresorufin O-debenzylase (BROD) as CYP2B1-dependent monooxygenase 3.6- and 4.1-fold in rats. On the contrary, these enzyme activities in mice were induced by CNP rather than by CNP-amino. Furthermore, immunoblotting showed that the protein levels of CYP2B subfamily cytochrome P450 (P450) in liver microsomes of rats and mice were increased by CNP or CNP-amino. Phase II drug-metabolizing enzymes, UDP-glucuronyltransferase (UGT) and glutathione S-transferase (GST) levels in mice were also significantly increased from 1.4 to 2.5-fold by CNP or CNP-amino. However, neither CNP nor CNP-amino affected UGT and GST in rats. These results suggest that CNP and or CNP-amino induce the P450 isoforms of CYP2B subfamily in the rat and mouse liver, and that the inducibility of drug-metabolizing enzyme by the compounds is different between rats and mice.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Chlornitrofen, PESTANAL®, analytical standard