Merck
  • Stimulatory effects of bombesin-like peptides on hypothalamic arcuate neurons in rat brain slices.
Stimulatory effects of bombesin-like peptides on hypothalamic arcuate neurons in rat brain slices.
Brain research bulletin (1994-01-01)
J Y Lin, J T Pan
ZUSAMMENFASSUNG
The effects of bombesin, gastrin-releasing peptide, neuromedin C, ranatensin, and neuromedin B on hypothalamic arcuate neurons were tested in this study using extracellular single-unit recording in fresh brain tissue slices. Adult ovariectomized Sprague-Dawley rats were used for preparation of brain slices. All bombesin-like peptides in pmol ranges exhibited potent stimulatory effects on the firing of arcuate neurons, i.e., gastrin-releasing peptide stimulated 90.9% (n = 22), bombesin 78.0% (n = 41), neuromedin C 63.2% (n = 19), ranatensin 58.0% (n = 22), and neuromedin B 50.0% (n = 6) of arcuate neurons tested. Pretreatments with either [Leu13-psi(CH2NH)-Leu14]-bombesin or [D-Phe6,Des-Met14]-bombesin6-14 ethylamide, two bombesin antagonists, significantly blocked most of the actions of bombesin-like peptides tested. The present results further support the notion that bombesin-like peptides may play a significant role in the arcuate nucleus.