Merck
  • Rapid assay of the anti-inflammatory activity of topical corticosteroids by inhibition of a UVA-induced neutrophil infiltration in hairless mouse skin. II. Assessment of name brand versus generic potency.
Rapid assay of the anti-inflammatory activity of topical corticosteroids by inhibition of a UVA-induced neutrophil infiltration in hairless mouse skin. II. Assessment of name brand versus generic potency.
Acta dermato-venereologica (1994-01-01)
L H Kligman
ZUSAMMENFASSUNG
The hairless mouse model of a UVA-induced dermal neutrophilic infiltrate was used to compare the efficacy of equal concentrations of name brand versus generic corticosteroids. The generic brand was significantly less effective in suppressing the inflammatory response.