Merck
Antagonists of 5-HT4 receptor-mediated responses in adult hippocampal neurons.
The Journal of pharmacology and experimental therapeutics (1994-10-01)
G E Torres, I L Holt, R Andrade
ZUSAMMENFASSUNG
The study of serotonin-4 (5-HT4) receptors in the central nervous system has been hindered by the lack of effective, selective antagonists. However, recently, several novel compounds have been synthesized and shown to act as antagonists at 5-HT4 receptors in smooth muscle and embryonic neurons in culture. In the present study, intracellular electrophysiological recordings were used to test the effects of three of these compounds: endo-8-methyl-8-azabicyclo[3.2.1]oct-3-yl-2,3-dihydro-6-methoxy- 2-oxo-1H-benzimidazole-1-carboxylate (DAU 6285), [1-[2-(methylsulfonylamino)ethyl]-4-piperidinyl]methyl 1-methyl-1H-indole-3-carboxylate (GR 113808) and 2-diethylaminoethyl-(2-methoxy-4-amino-5-chloro) benzoate (SDZ 205-557) on the 5-HT4 reduction of the afterhyperpolarization seen in adult CA1 hippocampal neurons in brain slices. GR 113808, SDZ 205-557 and DAU 6285 all functioned as competitive antagonists at these 5-HT4 receptors. Although all three compounds tested acted as effective antagonists, they differed considerably in potency. When the potency of these antagonists at the 5-HT4 receptor that mediates the reduction of the afterhyperpolarization was compared with that observed for 5-HT4 receptors in biochemical and binding assays, an excellent correlation was observed. Among the antagonists tested, GR 113808 was the most potent (pA2 = GR 113808 > SDZ 205-507 > DAU 6285). It exhibited an apparent affinity for the 5-HT4 receptors in the low nanomolar range but did not antagonize 5-HT1A, beta-adrenergic or muscarinic receptor-mediated responses when applied at concentrations two orders of magnitude higher.(ABSTRACT TRUNCATED AT 250 WORDS)