Merck
  • Pharmacokinetic and toxicological aspects of the medication of beef-type calves with an oral formulation of chloramphenicol palmitate.
Pharmacokinetic and toxicological aspects of the medication of beef-type calves with an oral formulation of chloramphenicol palmitate.
Journal of veterinary pharmacology and therapeutics (1994-08-01)
B Gassner, A Wuethrich
ZUSAMMENFASSUNG
Chloramphenicol (CAP) plasma levels were determined after oral administration of four doses of CAP palmitate (each dose corresponding to CAP 25 mg/kg/12 h) to four ruminating beef-type calves. Steady-state plasma concentrations of CAP were reached after the fourth oral dose and varied between 5 and 6 micrograms/ml. Half-life of elimination of CAP was 4.5 h. In addition to CAP, dehydrochloramphenicol (DH-CAP), a metabolite of chloramphenicol, was detected in plasma at concentrations between 3 and 7 ng/ml. DH-CAP is known to be produced from CAP by intestinal bacteria. This is significant since DH-CAP is suspected of being involved in the development of fatal aplastic anaemia, which occurs in man after exposure to CAP. Thus, it cannot be excluded that DH-CAP residues may occur in edible tissues. A risk arising from DH-CAP can neither be excluded for the animals being treated with CAP nor for consumers.