- Thermal stabilities of hemoglobins crosslinked with different length reagents.
Thermal stabilities of hemoglobins crosslinked with different length reagents.
Artificial cells, blood substitutes, and immobilization biotechnology (1994-01-01)
H Huang, K W Olsen
PMID7994393
ZUSAMMENFASSUNG
Bis(3,5-dibromosalicyl) succinate and glutarate were used to crosslink met-, oxy- and deoxyhemoglobins. The added flexibility of these reagents compared to the fumarate analog resulted in a more heterogeneous product but did not greatly affect the maximum thermal stability of the crosslinked hemoglobins.