Merck
Food restriction and stimulation of monooxygenation of p-nitroanisole in perfused rat liver.
Biochemical pharmacology (1994-07-19)
W Qu, F C Kauffman, R G Thurman
ZUSAMMENFASSUNG
This study assessed the effect of food restriction on the metabolism of model monooxygenase substrates in the perfused rat liver. Female Sprague-Dawley rats has access ad lib. to a Purina 5001 nonpurified diet (control) or were given 65% of the intake of controls for 3 weeks. Livers were perfused with oxygenated Krebs-Henseleit buffer using a non-recirculating system, and the rates of monooxygenation of p-nitroanisole and 7-ethoxycoumarin were measured. The results indicate that food restriction stimulated p-nitroanisole O-demethylation from 2.9 +/- 0.2 to 4.6 +/- 0.5 mumol/(g.hr) when saturating concentrations of p-nitroanisole were infused. Concomitantly, the ratio of beta-hydroxybutyrate to acetoacetate (B/A) and the rates of ketogenesis (B + A) were increased significantly by food restriction. Further, p-nitroanisole (200 mumol/L) increased hepatic malate concentration nearly 3-fold in liver extracts from food-restricted rats. However, infusion of either a low concentration of p-nitroanisole (50 mumol/L) or 7-ethoxycoumarin (200 mumol/L) did not alter these parameters. On the other hand, food restriction did not alter rates of monooxygenation in isolated microsomes supplemented with excess NADPH. Taken together, these data support the hypothesis that high concentrations of p-nitroanisole increased monooxygenation in food-restricted rats by stimulating fatty acid oxidation, which elevates the mitochondrial NADH/NAD+ ratio. This, in turn, increases the availability of reducing equivalents in the form of NADPH by a malate-pyruvate exchange system, leading to increased drug metabolism.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Nitroanisol, 97%