Merck
  • Acute toxicity of nifurpirinol, a fish chemotherapeutant, to milkfish (Chanos chanos) fingerlings.
Acute toxicity of nifurpirinol, a fish chemotherapeutant, to milkfish (Chanos chanos) fingerlings.
Bulletin of environmental contamination and toxicology (1994-03-01)
C T Tamse, R Q Gacutan