Merck
  • Dephostatin, a novel protein tyrosine phosphatase inhibitor produced by Streptomyces. II. Structure determination.
Dephostatin, a novel protein tyrosine phosphatase inhibitor produced by Streptomyces. II. Structure determination.
The Journal of antibiotics (1993-11-01)
H Kakeya, M Imoto, Y Takahashi, H Naganawa, T Takeuchi, K Umezawa
ZUSAMMENFASSUNG
Dephostatin, a novel tyrosine phosphatase inhibitor, was isolated from the culture broth of Streptomyces sp. MJ742-NF5. The structure was elucidated to be 2-(N-methyl-N-nitroso)hydroquinone by spectral and chemical analyses of dephostatin and its derivatives.