Merck
Cushing's syndrome from topical glucocorticoids.
The West Indian medical journal (1993-06-01)
S Teelucksingh, M Bahall, D Coomansingh, M Suite, C Bartholomew
ZUSAMMENFASSUNG
A case of Cushing's syndrome is described in a woman who self-treated psoriasis with a variety of potent topical glucocorticoids over 15 years. She was successfully weaned off corticosteroids and was treated with alternative anti-psoriatic drugs. The disappearance, nine months later, of most features of Cushing's syndrome, and the normal suppression of cortisol in response to dexamethasone, excluded endogenous hypercorticolism. The apparent widespread availability across the counter of potent corticosteroids is a cause for concern.