Merck
The Cardiac Arrhythmia Suppression Trial: Implications for nursing practice.
American journal of critical care : an official publication, American Association of Critical-Care Nurses (1996-01-01)
J C Kellen, A Ettinger, L Todd, M L Brezsnyak, J Campion, R McBride, S Thomas, J Corum, E Schron
ZUSAMMENFASSUNG
Care of patients with ventricular arrhythmia after myocardial infarction requires careful nursing management, including assisting with arrhythmia monitoring and testing. Because ventricular premature depolarization is a known risk factor for sudden cardiac death, it was hypothesized that the suppression of asymptomatic or mildly symptomatic ventricular premature depolarization would improve survival in these patients. To review the Cardiac Arrhythmia Suppression Trial findings and provide implications for nursing practice for patients after myocardial infarction. The Cardiac Arrhythmia Suppression Trial was a multicenter, randomized, placebo-controlled trial designed to determine whether the suppression of ventricular premature depolarizations in postmyocardial infarction patients would improve survival. Three class I antiarrhythmic drugs were used: encainide, flecainide, or moricizine. Patients for whom the drug suppressed their arrhythmia 80% or more were randomly assigned to that drug and dose or its matching placebo and were followed every 4 months (main study). Patients with 1% to 79% suppression were randomly assigned to the drug or its placebo that best treated their arrhythmia and followed every 4 months. Suppression of asymptomatic or mildly symptomatic ventricular premature depolarization in patients using encainide, flecainide, or moricizine failed to improve patient survival and was even harmful in some cases. Our results showed that in the absence of effective antiarrhythmic drug therapy, supportive nursing care and arrhythmia monitoring is important until appropriate therapy for the management of these arrhythmias in patients who have had a myocardial infarction can be found. Clinical trials are essential to provide an evaluation of therapies and direction for further studies, as well as a basis for practicing clinicians.