Merck
  • Carbachol stimulates inositol phosphate formation in rat thalamus slices through muscarinic M3-receptor activation.
Carbachol stimulates inositol phosphate formation in rat thalamus slices through muscarinic M3-receptor activation.
Neuroscience letters (1996-07-26)
M T de la Vega, A Nunez, J A Montano
ZUSAMMENFASSUNG
In cross-chopped slices from rat thalamus and in the presence of 10 mM LiC1, the cholinergic agonist carbachol stimulated the accumulation of total [3H]inositol phosphates ([3H]IP2 = [3H]IP1 + [3H]IP2 + [3H]IP3). Best-fit values for the concentration-response curve for carbachol after 60 min incubation yielded an EC50 of 44 +/- 6 microM, maximum effect of 199 +/- 6% of basal accumulation and Hill coefficient (nH) of 1.1 +/- 0.1. Carbachol-induced [3H]IPs accumulation was inhibited by 4-diphenylacetoxy-N-methylpiperidine methiodide (4-DAMP; pKi 9.1) and the p-fluoro analogue of hexahydro-sila-difenidol (pF-HHSiD; pKi 8.1). Concentration-response curves for carbachol were shifted to the right in a parallel fashion by pirenzepine (100, 300 and 100 nM). A Schild plot of the data was linear (slope 0.95 +/- 0.04) and yielded a log KD for pirenzepine of -6.8 +/- 0.1. Taken together, these results suggest that carbachol-induced inositol phosphate accumulation in rat thalamus is mediated by muscarinic M3-receptors.