Merck
  • Determination of antifungals in rodent diet by supercritical fluid extraction followed by packed column supercritical fluid chromatography with ultraviolet detection.
Determination of antifungals in rodent diet by supercritical fluid extraction followed by packed column supercritical fluid chromatography with ultraviolet detection.
Journal of AOAC International (1997-01-01)
J R Dean, I A Fowlis, S M Hitchen, S Khundker, E Ludkin, F Normand, P Jones
ZUSAMMENFASSUNG
Supercritical fluid extraction (SFE) followed by packed column supercritical fluid chromatography with ultraviolet detection was evaluated as a quantitative method for determining 4 antifungals (fluconazole, tioconazole, hexaconazole, and UK-47,265) in rodent diet. Chromatography was achieved with a cyano-bonded silica column, UV detection at 210 nm, and methanol-modified supercritical carbon dioxide as mobile phase. The effects of modifier concentration, temperature, and column pressure on antifungal retention time was studied. Off-line SFE was optimized at 2 spike levels, ranging from 0.5 to 10 g/kg, for each of the 4 antifungals. Average recoveries ranged from 79.0% for UK-47,265 to 96.5% for hexaconazole. Overall, the procedure provides a suitable method for analyzing antifungals in spiked rodent diet.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tioconazol, 97%