Merck
Antioxidant functions of inositol 1,2,3-trisphosphate and inositol 1,2,3,6-tetrakisphosphate.
Free radical biology & medicine (1997-01-01)
B Q Phillippy, E Graf
ZUSAMMENFASSUNG
Iron chelates of inositol 1,2,3-trisphosphate and inositol 1,2,3,6-tetrakisphosphate lacked free coordination sites and prevented the iron-catalyzed oxidation of ascorbic acid and peroxidation of arachidonic acid. In contrast, iron chelates of inositol 1,2,6-trisphosphate and inositol 1,2,5,6-tetrakisphosphate contained available coordination sites, permitted iron-catalyzed ascorbic acid oxidation, and enhanced arachidonic acid peroxidation. It was concluded that the 1,2,3-trisphosphate grouping of inositol hexakisphosphate was responsible for the inhibition of iron-catalyzed hydroxyl radical formation. The structure of the chelate with the phosphates in an axial-equatorial-axial configuration appeared to be the only possible inositol trisphosphate that could form bonds between six oxygen atoms and the six coordination sites on iron. Km values for cleavage by Escherichia coli alkaline phosphatase were as follows: inositol 1,2,3-trisphosphate, 56 microM; inositol 1,2,6-trisphosphate, 35 microM; inositol 1,2,3,6-tetrakisphosphate, 139 microM; and inositol 1,2,5,6-tetrakisphosphate, 100 microM. The initial hydrolysis rates of 200 microM solutions of the latter three isomers by E. coli alkaline phosphatase were not affected by an equimolar concentration of iron, whereas the rate for inositol 1,2,3-trisphosphate decreased in the presence of iron to 50% of the control. Therefore, the antioxidant potential of inositol 1,2,3-trisphosphate and inositol 1,2,3,6-tetrakisphosphate in cells and other biological systems may be fortified by the resistance of their iron chelates to enzymatic hydrolysis of the functional 1,2,3-trisphosphate array.