Merck
  • [The characteristics of extracting 4-methyl-2,6-dinitrophenol and 2-(1-methylpropyl)-4,6-dinitrophenol from aqueous solutions].
[The characteristics of extracting 4-methyl-2,6-dinitrophenol and 2-(1-methylpropyl)-4,6-dinitrophenol from aqueous solutions].
Sudebno-meditsinskaia ekspertiza (1997-01-01)
V K Shormanov, N G Denisova