Merck
  • New method for assaying free and total cholesterol in cultured cells by high-pressure liquid chromatography.
New method for assaying free and total cholesterol in cultured cells by high-pressure liquid chromatography.
Journal of atherosclerosis and thrombosis (1996-01-01)
K Hayashi, Y Kuga, Y Okura, K Tanaka, Y Yasunobu, K Nomura, Y Toyota, H Ohtani, T Shingu, G Kajiyama
ZUSAMMENFASSUNG
We developed a simple, sensitive and accurate method for assaying cellular free and total cholesterol by monitoring 4-cholesten-3-one, a conversion product of the cholesterol oxidase-catalyzed oxidation of the free cholesterol that has a strong chromophoric alpha, beta-unsaturated ketone at 240 nm, using a high-pressure liquid chromatographic system. This method measured picomole quantities of free and total cholesterol and precisely determined their concentrations in cells (10(4) range) in culture using 7 beta-hydroxycholesterol as an internal standard.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(+)-4-Cholesten-3-on, 98%