Merck
  • Measurements of urinary prostaglandins in young ovulatory women during the menstrual cycle and in postmenopausal women.
Measurements of urinary prostaglandins in young ovulatory women during the menstrual cycle and in postmenopausal women.
Prostaglandins (1997-11-28)
K Farker, H Schweer, R Vollandt, N Nassr, U Nagel, H W Seyberth, A Hoffmann, M Oettel
ZUSAMMENFASSUNG
The purpose of the present work was to study the prostaglandin excretion in young nonpregnant ovulatory women during the menstrual cycle on the one hand and in postmenopausal women on the other hand and to investigate the influence of female sex hormones (estradiol, progesterone) on urinary prostanoid excretion. Urinary excretion rates of prostaglandin E2 (PGE2), 6-keto-PGF1 alpha, thromboxane B2 (TxB2) and their metabolites PGE-M (11 alpha-hydroxy-9, 15-dioxo-2,3,4,5,20-pentanor-19-carboxyprostanoic acid), 2,3-dinor-6-keto-PGF1 alpha, 2,3-dinor-TxB2 and 11-dehydro-TxB2 were determined by gas chromatography-triple stage quadrupole mass spectrometry (GC/MS/MS) in 41 young non-pregnant women during the follicular phase and during the luteal phase and in 23 postmenopausal women. Excretion rates of all urinary prostanoids were not significantly different in the follicular phase when compared with the luteal phase. In contrast to the young ovulatory women, PGE2 and TxB2 were significantly higher in postmenopausal women. Concerning the other prostaglandins significant differences between these groups of women did not exist. Although serum levels of estradiol and progesterone were different in young and postmenopausal women, sex hormones have not been shown to correlate with prostaglandins. Our data do not suggest sex hormones to be responsible for the difference in the prostaglandin excretion in women of reproductive age and in women in the menopause. Further systematic investigations into age dependency of prostaglandin excretion in women are necessary.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Progesteron, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesteron, ≥99%
Sigma-Aldrich
Estron, ≥99%
Sigma-Aldrich
Progesteron, meets USP testing specifications
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesteron, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Supelco
Progesteron, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Androstenedione solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Progesterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Estrone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
4-Androsten-3,17-dion, VETRANAL®, analytical standard
Supelco
Progesteron, VETRANAL®, analytical standard
Dinoproston, European Pharmacopoeia (EP) Reference Standard
Estron, European Pharmacopoeia (EP) Reference Standard