- Mycotoxins of Alternaria alternata produced on ceiling tiles.
Mycotoxins of Alternaria alternata produced on ceiling tiles.
Journal of industrial microbiology & biotechnology (1998-04-02)
P Ren, D G Ahearn, S A Crow
PMID9523456
ZUSAMMENFASSUNG
The production of mycotoxins by Alternaria alternata in cellulosic ceiling tiles was examined with thin-layer chromatography and high-performance liquid chromatography procedures. Alternariol and alternariol monomethyl ether were found in ceiling tile extracts, whereas extracts of control rice cultures of all three isolates produced these mycotoxins plus altenuene and altertoxin I. Extensive fungal growth and mycotoxin production occurred in the ceiling tiles at relative humidities of 84-89% and 97%.