Merck
  • Activation of neurotensin receptors in the prefrontal cortex stimulates midbrain dopamine cell firing.
Activation of neurotensin receptors in the prefrontal cortex stimulates midbrain dopamine cell firing.
European journal of pharmacology (1998-05-23)
P P Rompré, S M Boye, J Moisan
ZUSAMMENFASSUNG
The effects of medial prefrontal cortex microinjections of 3 nmol/0.5 microl of neurotensin-(1-13), the inactive fragment neurotensin-(1-8), or vehicle on the firing rate of midbrain dopamine neurons were studied in anesthetized rats. Twelve of 19 cells tested with neurotensin-(1-13) showed an average 20-25% increase in firing rate between 10 and 20 min after the injection. This effect was not mimicked by neurotensin-(1-8) (9 cells), nor by a control injection (10 cells) suggesting that it is mediated by high-affinity neurotensin receptors. These results suggest that activation of neurotensin receptors in the medial prefrontal cortex can modulate neural activity of a subpopulation of midbrain dopamine neurons.