Merck
  • Synthesis of 4 alpha-(2-propenyl)-5,6-secocholestan-3 alpha-ol, a novel B-ring seco analog of the hypocholesterolemic agent 4 alpha-(2-propenyl)-5 alpha-cholestan-3 alpha-ol.
Synthesis of 4 alpha-(2-propenyl)-5,6-secocholestan-3 alpha-ol, a novel B-ring seco analog of the hypocholesterolemic agent 4 alpha-(2-propenyl)-5 alpha-cholestan-3 alpha-ol.
Steroids (1998-05-20)
H S Lin, A A Rampersaud, M E Richett, L S Beavers, D B McClure, R A Gadski
ZUSAMMENFASSUNG
4 alpha-(2-Propenyl)-5 alpha-cholestan-3 alpha-ol (LY295427) was previously identified from a CHO cell-based assay to be a potent LDL receptor up-regulator and had demonstrated to be an effective agent in lowering plasma cholesterol levels in hypercholesterolemic hamsters. In order to investigate the effect of flexibility of the 3 alpha-hydroxy-bearing A-ring on the activity, 4 alpha-(2-propenyl)-5,6-secocholestan-3 alpha-ol (11), a B-ring seco analog of LY295427, is thus synthesized from cholest-4-en-3-one. Test results indicate that 11 is not active in the CHO cell-based LDL receptor/luciferase assay at concentrations up to 20 micrograms/mL. The result underlines the importance of maintaining the A-B-C-D ring rigidity of the 3 alpha-sterols in terms of binding to the putative oxysterol receptor.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(+)-4-Cholesten-3-on, 98%