Merck
  • Quantitative determination of acrivastine and pseudoephedrine hydrochloride in pharmaceutical formulation by high performance liquid chromatography and derivative spectrophotometry.
Quantitative determination of acrivastine and pseudoephedrine hydrochloride in pharmaceutical formulation by high performance liquid chromatography and derivative spectrophotometry.
Journal of pharmaceutical and biomedical analysis (1998-06-03)
T G Altuntas, S S Zanooz, D Nebioglu
ZUSAMMENFASSUNG
In this study, fourth derivative spectrophotometry and high performance liquid chromatography (HPLC) have been used and described for the quantitative determination of acrivastine (I) and pseudoephedrine hydrochloride (II) in their pharmaceutical capsules form (Duact). In the former method, d4A/d gamma 4 values were measured in methanol at 315 and 269 nm for (I) and (II) respectively. The relative standard deviations (RSD) for the method were found to be 1.16% for (I) and 0.94% for (II). The latter method based on reversed phase HPLC system using LiChrosorb C18 analytical column. The mobile phase used for separation of (I), (II) and internal standard (p-hydroxymethylbenzoate) were the water/acetonitrile/methanol/perchloric acid/n-octylamine (500:130:25:13:0.3 v/v) and the detection of the compounds in the capsules were at 260 nm using UV detector. The RSD for the HPLC method were determined to be 0.79 and 0.88% for (I) and (II) respectively. The proposed methods, which give thoroughly comparable data, are simple, rapid, and allow precise and accurate results and could be used for commercial formulations containing acrivastine and pseudoephedrine hydrochloride in combination.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acrivastine, ≥98% (HPLC)